Check Order Status
Mineral Oil, Light Weight, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Mineral Oil, Light Weight, MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219483690
500 mL
Catalog number
MPB0219483690
Price (SGD)
97.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
97.00
EA
Add to cart
Specifications
Chemical Name or Material
Mineral Oil
CAS
8042-47-5
Recommended Storage
Store at room temperature.
Density
0.81 to 0.894g/mL
EINECS Number
232-455-8
View more