Check Order Status
Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate, ≥99%, Molecular Biology Reagent Grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate, ≥99%, Molecular Biology Reagent Grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219482280
100 g
Catalog number
MPB0219482280
Price (SGD)
99.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
99.00
EA
Add to cart
Specifications
Chemical Name or Material
Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate
CAS
6381-92-6
Color
White
Recommended Storage
Store at Room Temperature (15 to 30°C).
EINECS Number
205-358-3
View more