Check Order Status
DL-Dithiothreitol, 99.5%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
DL-Dithiothreitol, 99.5%, MP Biomedicals™
Have Questions?
Change view
Quantity:
25 g
5 g
10 g
Catalog number
MPB0219482125
Price (SGD)
1,134.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
1,134.00
EA
Add to cart
Specifications
Chemical Name or Material
DL-Dithiothreitol
CAS
27565-41-9
Recommended Storage
Store at + 4°C
EINECS Number
248-531-9
Assay Percent Range
>99%
View more