D-(+)-Galactose Anhydrous, cell culture reagent, 98.5%, MP Biomedicals™
D-(+)-Galactose Anhydrous, cell culture reagent, 98.5%, MP Biomedicals™
D-(+)-Galactose Anhydrous, cell culture reagent, 98.5%, MP Biomedicals™
D-(+)-Galactose Anhydrous, cell culture reagent, 98.5%, MP Biomedicals™
D-(+)-Galactose Anhydrous, cell culture reagent, 98.5%, MP Biomedicals™
MP Biomedicals
D-(+)-Galactose Anhydrous, cell culture reagent, 98.5%, MP Biomedicals™
Have Questions?
Catalog NumberQuantity
MPB0219467090500 g
Catalog number MPB0219467090
Price (SGD)
463.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
463.00
EA
Add to cart
Specifications
CAS10257-28-0
Molecular FormulaC6H12O6
Quantity500 g
Synonymd-galactose, d-galactopyranose, galactose, galactopyranose, galactopyranoside, d-+-galactose, galactose, pure, cerebrose, gal, d-galactopyranoside
InChI KeyWQZGKKKJIJFFOK-UHFFFAOYNA-N
View more