Citric Acid, MP Biomedicals™
Citric Acid, MP Biomedicals™
MP Biomedicals
Citric Acid, MP Biomedicals™
Citric Acid acts as an iron chelator.
Have Questions?
Catalog NumberQuantity
MPB0219464180100 g
Catalog number MPB0219464180
Price (SGD)
73.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
73.00
EA
Add to cart
Specifications
pH1 to 3 (5% aq. soln.)
CAS77-92-9
Melting Point149°C to 159°C
Molecular FormulaC6H8O7
Solubility InformationSoluble in water (50 g/mL)
View more
  • Citric acid can support cholesterol/sterol, ubiquinone and isoprenoid biosynthesis in cell culture applications.
  • In animals, citric acid improves the utilization of nutritional calcium.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.