Check Order Status
D-Biotin, ≥97.5%, Cell Culture Reagent Grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
D-Biotin, ≥97.5%, Cell Culture Reagent Grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219463401
1 g
Catalog number
MPB0219463401
Price (SGD)
294.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (SGD)
294.00
EA
Add to cart
Specifications
Chemical Name or Material
D-Biotin
CAS
58-85-5
Color
White
Recommended Storage
Store at +4°C.
Density
1.267 g/cm
3
(Lit.)
View more