Penicillin G, Sodium Salt, MP Biomedicals™
Penicillin G, Sodium Salt, MP Biomedicals™
Penicillin G, Sodium Salt, MP Biomedicals™
MP Biomedicals
Penicillin G, Sodium Salt, MP Biomedicals™
Penicillin G, Sodium Salt Cell culture grade, Powder
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mU
100 mU
Catalog number MPB0219453725
Price (SGD)
183.00
Each
Quantity:
25 mU
Request bulk or custom format
Price (SGD)
183.00
Each
Specifications
Physical FormPowder
CAS69-57-8
Storage Note 1Store at +4°C, protect from light.
Loss on Drying0.001
RTECS NumberXH9800000
View more
  • Penicillin G is a narrow spectrum antibiotic derived from Streptococcus pneumoniae.
  • It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility and in cell culture both alone and combined with streptomycin and other antibiotics.
  • Penicillin G also known as benzylpenicillin.
  • It exert a bacteriocidal as well as a bacteriostatic effect on susceptible bacteria by interfering with the final stages of the synthesis of peptidoglycan, a substance in the bacterial cell wall.