Check Order Status
Colistin, sulfate salt, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Colistin, sulfate salt, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219415791
1 g
Catalog number
MPB0219415791
Price (SGD)
376.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (SGD)
376.00
EA
Add to cart
Specifications
Chemical Name or Material
Colistin, sulfate salt
CAS
1264-72-8
Color
Brown
Quantity
1 g
Physical Form
Powder
View more