▸
Solvents
▸
Chloroform
▸
Chloroform Molecular Biology MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Chloroform Molecular Biology MP Biomedicals
Have Questions?
Change view
Quantity:
0.5 L
1 L
Catalog number
MPB0219400290
Price (SGD)
134.00
Each
Add to cart
Quantity:
0.5 L
Request bulk or custom format
Price (SGD)
134.00
Each
Add to cart
Specifications
Chemical Name or Material
Chloroform
CAS
67-66-3
Density
4.36 (air = 1)
Quantity
0.5 L
Molecular Formula
CHCl3
View more