Check Order Status
ChloroForm, Reagent grade, For ACS analysis, ≥99.8%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
ChloroForm, Reagent grade, For ACS analysis, ≥99.8%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219381490
0.5 L
Catalog number
MPB0219381490
Price (SGD)
107.00
EA
Add to cart
Quantity:
0.5 L
Request bulk or custom format
Price (SGD)
107.00
EA
Add to cart
Specifications
Chemical Name or Material
Chloroform
Name Note
For ACS analysis
CAS
67-66-3
Density
1.476 to 1.478 g/cm
3
at 20°C
Quantity
0.5 L
View more