Check Order Status
Potassium Iodide, (Crystalline),MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Potassium Iodide, (Crystalline),MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219151380
100 g
Catalog number
MPB0219151380
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material
Potassium Iodide
CAS
7681-11-0
Recommended Storage
Room Temperature
Physical Form
Crystalline
Quantity
100 g
View more