Check Order Status
Sodium Phosphate Dibasic, (White, crystalline powder/ACS Reagent Grade), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Sodium Phosphate Dibasic, (White, crystalline powder/ACS Reagent Grade), MP Biomedicals
Have Questions?
Catalog number
MPB0219144001
Price (SGD)
189.00
Each
Add to cart
Request bulk or custom format
Price (SGD)
189.00
Each
Add to cart
Specifications
Chemical Name or Material
Sodium Phosphate Dibasic
CAS
7558-79-4
Color
White
Physical Form
Crystalline Powder
Quantity
1 kg
View more