Sodium Phosphate Dibasic, MP Biomedicals™
Sodium Phosphate Dibasic, MP Biomedicals™
Sodium Phosphate Dibasic, MP Biomedicals™
Sodium Phosphate Dibasic, MP Biomedicals™
MP Biomedicals
Sodium Phosphate Dibasic, MP Biomedicals™
Widely used in molecular biology, analytical chemistry, biochemistry and chromatography. Sodium Phosphate Dibasic, MP Biomedicals is a reagent with very high buffering capacity, often used in conjunction with Sodium Phosphate, Monobasic in the preparation
Have Questions?
Catalog NumberQuantity
MPB02191440.5500 g
Catalog number MPB02191440.5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
CAS7558-79-4
Recommended StorageRoom Temperature (15-30°C), dessicate.
Physical FormCrystalline Powder
Quantity500 g
Molecular FormulaHNa2O4P
View more
Sodium Phosphate Dibasic is used in many applications, including the purification of antibodies, as a reagent and buffer in analytical chemistry, and in the preparation of biological buffers. Also used as a sequestrant, emulsifier and buffer in foods; as mordant in dyeing; for weighting silk; in tanning; in manufacturing of enamels, ceramics, detergents, and boiler compounds; as a fireproofing agent in soldering and brazing instead of borax; as a cathartic; and as a laxative.