Potassium Phosphate Monobasic Anhydrous, MP Biomedicals
Potassium Phosphate Monobasic Anhydrous, MP Biomedicals
Potassium Phosphate Monobasic Anhydrous, MP Biomedicals
Potassium Phosphate Monobasic Anhydrous, MP Biomedicals
MP Biomedicals
Potassium Phosphate Monobasic Anhydrous, MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB02191430.1100 g
Catalog number MPB02191430.1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
CAS7778-77-0
Quantity100 g
Molecular FormulaH2KO4P
MDL NumberMFCD00011401 MFCD00147253
Synonympotassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
View more