Check Order Status
Aluminum Sulfate Hydrate ACS MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Aluminum Sulfate Hydrate ACS MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219140390
500 g
Catalog number
MPB0219140390
Price (SGD)
74.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
74.00
Each
Add to cart
Specifications
Chemical Name or Material
Aluminum Sulfate Hydrate
Name Note
Hydrate
CAS
17927-65-0
Recommended Storage
Store at room temperature.
Quantity
500 g
View more