Check Order Status
Coomassie Brilliant Blue R-250, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Coomassie Brilliant Blue R-250, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219068225
25 g
Catalog number
MPB0219068225
Price (SGD)
147.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
147.00
EA
Add to cart
Specifications
Chemical Name or Material
Coomassie Brilliant Blue R-250
Name Note
Protein stain in electrophoresis.
CAS
6104-59-2
Assay Percent Range
∽ 50%
Molecular Formula
C45H44N3NaO7S2
View more