Ponceau S, 85%, MP Biomedicals Inc
Ponceau S, 85%, MP Biomedicals Inc
Ponceau S, 85%, MP Biomedicals Inc
Ponceau S, 85%, MP Biomedicals Inc
MP Biomedicals
Ponceau S, 85%, MP Biomedicals Inc
Have Questions?
Catalog NumberQuantity
MPB021906441010 g
Catalog number MPB0219064410
Price (SGD)
71.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (SGD)
71.00
EA
Add to cart
Specifications
Chemical Name or MaterialPonceau S
Name NoteProtein stain for electrophoresis
CAS6226-79-5
Assay Percent Range∽ 85%
Molecular FormulaC22H12N4Na4O13S4
View more