Check Order Status
Sodium dodecyl sulfate, 95%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Sodium dodecyl sulfate, 95%, MP Biomedicals™
Approximately 95% alkyl sulfate, predominantly composed of C
12
and C
14
homologs
Have Questions?
Catalog Number
Quantity
MPB0219052280
100 g
Catalog number
MPB0219052280
Price (SGD)
71.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
71.00
EA
Add to cart
Specifications
Chemical Name or Material
Sodium dodecyl sulfate
Color
White
CAS
151-21-3
Physical Form
Granular Powder
Density
>1.1 at 20°C (water = 1)
View more