Check Order Status
PIPES, 99.4%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
PIPES, 99.4%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219025790
500 g
Catalog number
MPB0219025790
Price (SGD)
671.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
671.00
EA
Add to cart
Specifications
Chemical Name or Material
Pipes
CAS
5625-37-6
Color
White
Physical Form
Crystalline Powder
Quantity
500 g
View more