▸
Cell Culture Media, Supplements, and Reagents
▸
Cell Culture Reagents, Antibiotics and Supplements
▸
Antibiotics
▸
Ampicillin Sodium Salt, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Ampicillin Sodium Salt, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219014805
5 g
Catalog number
MPB0219014805
Price (SGD)
125.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
125.00
Each
Add to cart
Specifications
Chemical Name or Material
Ampicillin sodium salt
Type
Ampicillin Sodium Salt
Physical Form
Powder
CAS
69-52-3
Quantity
5 g
View more