Check Order Status
Dextran sulfate sodium salt (400,000-600,000 M.Wt), MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Dextran sulfate sodium salt (400,000-600,000 M.Wt), MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0216011180
100 g
Catalog number
MPB0216011180
Price (SGD)
621.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
621.00
EA
Add to cart
Specifications
Chemical Name or Material
Dextran Sulfate Sodium Salt
Color
White
CAS
9011-18-1
Physical Form
Powder
Molecular Formula
(C6H7O5)m(C6H7O5)n
View more