▸
Organic compounds
▸
Organic Polymers
▸
Dextran sulfate sodium salt (36,000-50,000 M.Wt), Colitis Grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Dextran sulfate sodium salt (36,000-50,000 M.Wt), Colitis Grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0216011090
500 g
Catalog number
MPB0216011090
Price (SGD)
8,578.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
8,578.00
Each
Add to cart
Specifications
Chemical Name or Material
Dextran Sulfate Sodium Salt
Color
White
pH
5 to 7.10 (in 1% aq. soln.)
CAS
9011-18-1
Physical Form
Powder
View more
Dextran Sulfate Sodium (DSS) which has proven to have versatile value in Biomedical/clinical research.
DSS can be used to study Colitis (Inflammatory Bowel disease) as well as an antiviral as agent.
Dextran sulfate is a polyanionic derivative of dextran produced by esterification of Dextran with chlorosulphonic acid.
For research or further manufacturing use only, not for direct human use.