Permeabilizes plasma membranes of eukaryotic cells. Digitonin, MP Biomedicals is a mild nonionic detergent that complexes with membrane cholesterol and other unconjugated β-hydroxysterols. It is used to solubilize receptors and permeabilize cellular
Have Questions?
Catalog Number
Quantity
MPB0215948091
1 g
Catalog number MPB0215948091
Price (SGD)
2,756.00
Each
Quantity:
1 g
Request bulk or custom format
Price (SGD)
2,756.00
Each
Specifications
CAS11024-24-1
Physical FormWhite Powder
Melting Point230°C to 240°C (decomposition)(literature)
Recommended StorageStore at Room Temperature (15°C to 30°C), dessicate.
Quantity1 g
View more
Digitonin has following uses:
Cholesterol determination
Use of digitonin-permeabilized adipocytes for cAMP studies
Solubilization of membrane proteins
The water soluble form can be used for the isolation of mitotic chromosomes