Check Order Status
Ferrous Sulfate Heptahydrate MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Ferrous Sulfate Heptahydrate MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0215804891
1 kg
Catalog number
MPB0215804891
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or Material
Ferrous Sulfate
Name Note
Heptahydrate
CAS
7782-63-0
Quantity
1 kg
Molecular Formula
FeH14O11S
View more