Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Biochemical Reagents
▸
Peptides
▸
Phe-Arg-β-naphthylamide dihydrochloride, >99%, For TLC analysis, 85.4%, MP Biomedicals™
Actual product may vary
MP Biomedicals
Phe-Arg-β-naphthylamide dihydrochloride, >99%, For TLC analysis, 85.4%, MP Biomedicals™
Have Questions?
Change view
Catalog Number
Quantity
MPB0215612850
50mg
MPB0215612825
25mg
2 Options
Catalog number
MPB0215612850
Price (SGD)
491.00
Each
Add to cart
Quantity:
50mg
Price (SGD)
491.00
Each
Add to cart
Specifications
Purity
>99%
Assay
0.854
CAS
100929-99-5
Color
White
Format
Powder
Molecular Weight (g/mol)
519.471
Product Type
Phe-Arg-β-naphthylamide dihydrochloride
Quantity
50mg
Unit Size
Each
Contents & Storage
Store at 0°C.