Check Order Status
Nisin
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
MP Biomedicals
Nisin
Nisin content: 2.5% Remainder is NaCl and milk solids (denatured)
Have Questions?
Contact Us
Catalog Number
Quantity
MPB0215583905
5 g
Catalog number
MPB0215583905
Price (SGD)
285.00
EA
Estimated availability date
21-Sep-2025
Add to cart
Quantity:
5 g
Price (SGD)
285.00
EA
Add to cart
Molecular Formula: C
14
H
230
N
42
O
37
S
7
Molecular Weight: 3354.3
Specifications
Form
Powder
Product Type
Antimicrobial
pH
3.31
Quantity
5 g
Unit Size
EA
Contents & Storage
15° to 30°C
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
0 results displayed, search above for a specific certificate
Request a Certificate
Safety Data Sheets
Share catalog number, name or link