Check Order Status
D-Limonene, 96.9%, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
D-Limonene, 96.9%, MP Biomedicals
Have Questions?
Change view
Quantity:
500 mL
100 mL
Catalog number
MPB0215523490
Price (SGD)
155.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
155.00
EA
Add to cart
Specifications
Chemical Name or Material
D-Limonene
CAS
5989-27-5
Color
Colorless to Yellow
Molecular Formula
C10H16
MDL Number
MFCD00062991
View more