Cefotaxime Sodium Salt, MP Biomedicals™
Cefotaxime Sodium Salt, MP Biomedicals™
Cefotaxime Sodium Salt, MP Biomedicals™
MP Biomedicals
Cefotaxime Sodium Salt, MP Biomedicals™
Inhibits bacterial cell wall synthesis. Cefotaxime Sodium Salt, MP Biomedicals is a cephalosporin antibiotic that is active against both gram-negative and gram-positive bacteria. It is used for infections of the respiratory tract, skin, bones, joints, uro
Have Questions?
Catalog NumberQuantity
MPB02154947911 g
Catalog number MPB0215494791
Price (SGD)
435.00
Each
Quantity:
1 g
Request bulk or custom format
Price (SGD)
435.00
Each
Specifications
Physical FormPowder
CAS64485-93-4
Recommended StorageStore at 4°C, protect from light.
Quantity1 g
Molecular FormulaC16H16N5NaO7S2
View more
Cefotaxime binds to penicillin-binding proteins (PBPs), which inhibits the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls. As a result, bacteria lyse due to cell wall autolytic enzymes. Cefotaxime blocks the division of cyanobacteria and lower plant organelles/plastids such as the photosynthetic organelles of Glaucophytes and chloroplasts of bryophytes. Cefotaxime is used to treat a variety of bacterial infections:
  • Good coverage against most Gram-negative bacteria, with the notable exception of Pseudomonas
  • Effective against most Gram-positive cocci, except for Enterococcus
  • Active against penicillin-resistant strains of Streptococcus pneumoniae
  • Modest activity against the anaerobic Bacteroides fragilis
  • In meningitis, cefotaxime crosses the blood–brain barrier better than cefuroxime