Ferric Chloride Hexahydrate MP Biomedicals
Ferric Chloride Hexahydrate MP Biomedicals
Ferric Chloride Hexahydrate MP Biomedicals
MP Biomedicals
Ferric Chloride Hexahydrate MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB0215350080100 g
Catalog number MPB0215350080
Price (SGD)
58.00
Each
Quantity:
100 g
Request bulk or custom format
Price (SGD)
58.00
Each
Specifications
Chemical Name or MaterialFerric Chloride
Name NoteHexahydrate
CAS10025-77-1
Quantity100 g
Molecular FormulaCl3FeH12O6
View more