Check Order Status
Sodium Hydroxide, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Sodium Hydroxide, MP Biomedicals
Have Questions?
Change view
Quantity:
1 kg
500 g
Catalog number
MPB0215349501
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Hydroxide
Name Note
Anhydrous
CAS
1310-73-2
Recommended Storage
Room Temperature
Molecular Formula
HNaO
View more