▸
Biochemical Reagents
▸
Carbohydrates
▸
Monosaccharides
▸
D-(+)-Glucose, anhydrous, For ACS analysis, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
D-(+)-Glucose, anhydrous, For ACS analysis, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0215252790
500 g
Catalog number
MPB0215252790
Price (SGD)
82.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
82.00
Each
Add to cart
Specifications
Chemical Name or Material
D-(+)-Glucose
Name Note
For ACS analysis
CAS
50-99-7
Density
1.544g/mL
Molecular Formula
C6H12O6
View more