Check Order Status
Safranine O, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Safranine O, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0215204025
25 g
Catalog number
MPB0215204025
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material
Safranine O
Name Note
Stains and Dyes
CAS
477-73-6
Recommended Storage
Store at room temperature.
EINECS Number
207-518-8
View more