1x1 image pixel for data collection
Polyethylene Glycol, MP Biomedicals™
Polyethylene Glycol, MP Biomedicals™
Polyethylene Glycol, MP Biomedicals™
Polyethylene Glycol, MP Biomedicals™
MP Biomedicals
Polyethylene Glycol, MP Biomedicals™
Formed as a condensation polymer of ethylene oxide and water. Polyethylene Glycol, MP Biomedicals (PEG) has been used in many applications from medicine to industrial manufacturing, including the precipitation of proteins.
Have Questions?
Catalog NumberQuantity
MPB02151916.5500 g
Catalog number MPB02151916.5
Price (SGD)
208.00
EA
Estimated availability date 26-Jul-2025
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
208.00
EA
Add to cart
Specifications
CAS107-21-1
Melting Point55°C to 65°C
Recommended StorageRoom Temperature (15–30°C).
Density1.1–1.2g/mL (Lit.)
pH4.5 to 7.5 (5% aq. soln.)
View more
PEG has been used in a wide variety of biomedical and laboratory applications, including:
  • The precipitation of proteins
  • As a fusing agent in enhancing the effect of macrophages on hybridoma
  • In the separation and purification of biomolecules
  • In induction of cell hybridization
  • To recover pathogenic viruses and human rotaviruses from water, oyster and sediment samples
  • Demonstrated to have anticell proliferation and antitumor effects via induction of apoptosis
Utilization of PEG:
  • PEGs are susceptible to oxidative degradation in the presence of air; minimizing exposure to elevated temperatures and oxygen or addition of an antioxidant can limit the amount of degradation
  • PEGs do not hydrolyze or deteriorate upon storage
  • PEGs do not support the growth of molds
  • PEG is incompatible with phenol and may reduce the antimicrobial action of other preservatives
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link