Check Order Status
Phytohemagglutinin, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Phytohemagglutinin, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0215188401
1 mg
Catalog number
MPB0215188401
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 mg
Request bulk or custom format
Specifications
Chemical Name or Material
Phytohemagglutinin
Quantity
1 mg
Physical Form
Powder