Check Order Status
Glycerol, 99.91%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Glycerol, 99.91%, MP Biomedicals™
Have Questions?
Change view
Quantity:
1 L
500 mL
Catalog number
MPB0215119491
Price (SGD)
119.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
119.00
EA
Add to cart
Specifications
Chemical Name or Material
Glycerol
CAS
56-81-5
Color
Colorless
Density
1.2613g/cm
3
at 20°C
Assay Percent Range
0.9991
View more