Collagenase Type III from Clostridium histolyticum
Collagenase Type III from <i>Clostridium histolyticum</i>
Actual product may vary
MP Biomedicals
Collagenase Type III from Clostridium histolyticum
Collagenases degrades native helical collagen fibrils
Have Questions?
Catalog NumberQuantity
MPB02150704011 g
Catalog number MPB0215070401
Price (SGD)
949.00
EA
Add to cart
Quantity:
1 g
Price (SGD)
949.00
EA
Add to cart
  • CAS Number: 9001-12-1
  • EC Number: 232-582-9
  • MDL Number: MFCD00130830
  • Format: brown lyophilized powder
  • Specific Activity: >100U/mg
  • Optimum pH: 7 to 9
Collagenase has found widespread application in isolation of specific cell types from attendant connective tissue
Specifications
Concentration1 mg/mL
ColorBrown
pH7 to 9
For Use With (Application)Cell dissociation and cell lysis
Storage Buffer1mg/mL of Enzyme in 0.05M TES with 0.36mM Calcium Chloride, pH 7.5
SourceClostridium histolyticum
Quantity1 g
Product TypeCollagenase Type III
Molecular Weight (g/mol)105000 and 57400
InhibitorsMetal chelating agents such as Cysteine, EDTA or o-Phenanthroline
FormLyophilized Solid
Unit SizeEA
Contents & Storage
4°C