▸
Biochemical Reagents
▸
Carbohydrates
▸
Monosaccharides
▸
Carboxymethyl Cellulose Sodium Salt, Medium Viscosity, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Carboxymethyl Cellulose Sodium Salt, Medium Viscosity, MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0215056090
500 g
Catalog number
MPB0215056090
Price (SGD)
177.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
177.00
Each
Add to cart
Specifications
Chemical Name or Material
Carboxymethyl cellulose sodium salt
CAS
9004-32-4
Color
White
Recommended Storage
Store at Room Temperature (15 to 30°C).
Molecular Formula
(C12 H14 O9 R6)n
View more