Peptone from Porcine and/or Bovine Tissues, MP Biomedical
Peptone from porcine and/or bovine is Bacteriological Grade. It is a water-soluble product and is a mixture of polypeptides and amino acids formed by the partial hydrolysis of protein.
Have Questions?
Catalog Number
Quantity
MPB0210480801
1 lb.
Catalog number MPB0210480801
Price (SGD)
669.00
EA
Estimated availability date 21-Sep-2025
Add to cart
Quantity:
1 lb.
Request bulk or custom format
Price (SGD)
669.00
EA
Add to cart
Specifications
ColorTan
pH5.5 to 7.0 (10% aq. soln.)(literature)
PackagingPlastic
Quantity1 lb.
Unit SizeEA
Peptone is produced from porcine and/or bovine tissues
The tissues are digested enzymatically to create small peptides and then spray dried
Peptone is used in culture media in bacteriology
The growth requirements of most non fastidious organisms will be fulfilled by the range of amino acids, peptides, and proteases in this mixture