Product Image
MP Biomedicals
Casein Hydrolysate Acid Solution , MP Biomedicals, LLC
Vitamin free Casein hydrolysate acid solution is a hydrochloric acid digest of casein in which the protein molecule has been substantially reduced to its constituent amino acids
Have Questions?
Catalog NumberQuantityPackaging
MPB0210476990500 mLPlastic
Catalog number MPB0210476990
Price (SGD)
332.00
Each
Quantity:
500 mL
Packaging:
Plastic
Request bulk or custom format
Price (SGD)
332.00
Each
Specifications
ColorColorless
PackagingPlastic
Quantity500 mL
Concentration or Composition (by Analyte or Components)0.1
Unit SizeEach
Laboratory media including basal media for vitamin analysis of folic acid, pantothenic acid, riboflavin, thiamin, niacin, and B-12