Check Order Status
D(+)-Xylose, C.P. grade, 99% max., MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
D(+)-Xylose, C.P. grade, 99% max., MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210330080
100 g
Catalog number
MPB0210330080
Price (SGD)
69.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
69.00
EA
Add to cart
Specifications
Chemical Name or Material
D(+)-Xylose
CAS
58-86-6
Color
White
Recommended Storage
Store at Room Temperature (15° to 30°C)
EINECS Number
200-400-7
View more