Check Order Status
Tricine Biological buffer MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Tricine Biological buffer MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0210311291
1 kg
Catalog number
MPB0210311291
Price (SGD)
834.00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
834.00
Each
Add to cart
Specifications
Chemical Name or Material
Tricine
Name Note
Biological buffer
CAS
4-1-5704
Molecular Formula
C6H13NO5
Quantity
1 kg
View more