Check Order Status
Tetracyline hydrochloride, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Tetracyline hydrochloride, MP Biomedicals™
Have Questions?
Change view
Quantity:
25 g
5 g
Catalog number
MPB0210301125
Price (SGD)
114.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
114.00
Each
Add to cart
Specifications
Chemical Name or Material
Tetracyline hydrochloride
CAS
64-75-5
Color
Yellow
Molecular Formula
C22H25ClN2O8
Quantity
25 g
View more