Check Order Status
D-Sorbitol, >98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
D-Sorbitol, >98%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210293801
1 kg
Catalog number
MPB0210293801
Price (SGD)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or Material
D-Sorbitol
CAS
50-70-4
Color
White
Density
1.47g/mL at −5°C
Molecular Formula
C6H14O6
View more