L-(+)-Rhamnose, monohydrate, ∽99%, MP Biomedicals™
L-(+)-Rhamnose, monohydrate, ∽99%, MP Biomedicals™
L-(+)-Rhamnose, monohydrate, ∽99%, MP Biomedicals™
MP Biomedicals
L-(+)-Rhamnose, monohydrate, ∽99%, MP Biomedicals™
Have Questions?
Catalog NumberQuantity
MPB0210280980100 g
Catalog number MPB0210280980
Price (SGD)
608.00
Each
Quantity:
100 g
Request bulk or custom format
Price (SGD)
608.00
Each
Specifications
Chemical Name or MaterialL-(+)-Rhamnose, monohydrate
CAS10030-85-0
ColorWhite
Recommended StorageStore at room temperature.
Density1.47g/mL
View more