▸
Organic compounds
▸
Organic acids and derivatives
▸
Carboxylic acids and derivatives
▸
Monocarboxylic acids and derivatives
▸
n-Propyl Gallate, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
n-Propyl Gallate, MP Biomedicals™
Off-White Powder
Have Questions?
Catalog Number
Quantity
MPB0210274780
100 g
Catalog number
MPB0210274780
Price (SGD)
112.00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
112.00
Each
Add to cart
Specifications
CAS
121-79-9
Melting Point
140°C to 150°C
Storage Note 1
Store at Room Temperature (15-30°C).
Color
Off-white
RTECS Number
LW8400000
View more
Anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein
An ester formed by the condensation of gallic acid and propanol
This antioxidant has been added to foods containing oils and fats to prevent oxidation
An antioxidant that exhibits antimicrobial activity