▸
Organic compounds
▸
Organic Polymers
▸
Poly-D-Lysine Hydrobromide, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Poly-D-Lysine Hydrobromide, MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0210269480
100 mg
Catalog number
MPB0210269480
Price (SGD)
331.00
Each
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (SGD)
331.00
Each
Add to cart
Specifications
Chemical Name or Material
Poly-D-Lysine Hydrobromide
CAS
27964-99-4
Recommended Storage
0°C
Molecular Formula
C6H15BrN2O2
Synonym
d-lysine hydrobromide
View more