Check Order Status
Menadione, 98.76%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Menadione, 98.76%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210225980
100 g
Catalog number
MPB0210225980
Price (SGD)
327.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
327.00
EA
Add to cart
Specifications
Chemical Name or Material
Menadione
CAS
58-27-5
Color
Yellow
Molecular Formula
C11H8O2
Quantity
100 g
View more