Check Order Status
HEPES, free acid, >98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
HEPES, free acid, >98%, MP Biomedicals™
Have Questions?
Change view
Quantity:
1 kg
250 g
Catalog number
MPB0210192691
Price (SGD)
1,155.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
1,155.00
EA
Add to cart
Specifications
Chemical Name or Material
HEPES
CAS
7365-45-9
Color
White
Recommended Storage
Store at Room Temperature (15 to 30°C). Store Desiccated.
EINECS Number
230-907-9
View more