Check Order Status
Dextran Sulfate, Clinical grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Dextran Sulfate, Clinical grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210151880
100 g
Catalog number
MPB0210151880
Price (SGD)
1,919.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
1,919.00
EA
Add to cart
Specifications
Chemical Name or Material
Dextran Sulfate
Color
White Powder
CAS
9011-18-1
Physical Form
Solid
Recommended Storage
Store at Room Temperature (15deg to 30°C).
View more