▸
Organic compounds
▸
Organic Polymers
▸
Dextran sulfate sodium salt, Reagent grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Dextran sulfate sodium salt, Reagent grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0210151610
10 g
Catalog number
MPB0210151610
Price (SGD)
341.00
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (SGD)
341.00
Each
Add to cart
Specifications
Chemical Name or Material
Dextran sulfate sodium salt
Color
White
CAS
9011-18-1
Physical Form
Powder
Molecular Formula
(C6H7O5)m(C6H7O5)n
View more